Learn More
Chemsplash Overshoes (Pairs) 5 and 6, Anti-Slip Sole, Elasticated Ankle
GREENER_CHOICE
Product Code. p-200105900
Click to view available options
Size:
Medium
Large
X-Large
Unit Size:
200 pairs
Product Code. 30058349
Brand: Chemsplash 2564.400.005
Only null left
Add to basket
Add to basket
Anti-Slip Overshoe is made from Chemsplash fabric which is soft and microporous. It is strong and resistant to chemical splash and dust.
Chemsplash Anti-Slip Overshoe
  • Chemsplash 55GSM Type PB 6B Material
  • Coated White PVC Slip-Resistant Sole
  • Silicone and Latex Free
  • Anti-Static Material
  • Non-Linting
  • 16 in. in Length
  • Conforms to ISO 14644-1:2015 Cleanroom 5 - 9
  • PFAS Free
Description: The Chemsplash Anti-Slip Overshoe is made from Chemsplash fabric which is soft and microporous. It is strong and resistant to chemical splash and dust. It is anti-static to meet EN1149-5:2008, lint-free, and suitable for cleanroom applications. Conforms to ISO 14644-1:2015 Cleanroom 5 - 9. An anti-slip textured sole has been added to enhance grip with the floor, and an elasticated ankle ensures a secure fit. Chemsplash accessories can be used to protect the wearer from their environment or the environment from the wearer. The range has been designed to allow for maximum comfort and wearability.
Specifications
Size Medium
Material Microporous Fabric
Latex Free Yes
Color White
Disposable Single-use
Sterility Non-sterile
Closure Type Elastic Ankle
Sole Type Anti-slip
Resistance Liquid
Product Type Controlled Environments Shoe Covers
Cleanroom Class Class 100-1000 (ISO 5-6)