Learn More
Thermo Scientific™ Orion™ Single Junction Glass Bodied Combination Silver Billet Electrode
Product Code. 10429412
Change view
Click to view available options
Type:
Combination Electrode
Half Cell Electrode
Unit Size:
Each
Product Code. Type unitSize
10429412 Half Cell Electrode Each
10458912 Combination Electrode Each
2 options
Product Code. 10429412
Brand: Thermo Scientific™ 9781SC
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Receive an accurate determination of potentiometric halides with dual electrodes.
Thermo Scientific™ Orion™ Silver Billet Electrodes allow for an accurate determination of potentiometric halides. These electrodes are a combination of a silver billet-indicating electrode, along with a silver billet half-cell.
  • Available with combination electrode and half cell electrode feature
  • Concentration range: endpoint indicator
  • Temperature range: 0° to 80°C
  • Internal ref: Ag/AgCl
  • Dimensions: 120 × 12mm
TRUSTED_SUSTAINABILITY
Specifications
Description Silver half-cell construction
Diameter (Metric) Body 12 mm
Internal Reference Ag/AgCl
Length (Metric) 120 mm
Material Silver Billet
For Use With (Equipment) Orion instruments that perform potentiometric halide determinations
Temperature (Metric) Operating 0°C to 80°C
Type Half Cell Electrode