Learn More
Thermo Scientific™ Orion™ Redox/ORP/Temp Electrodes
Product Code. 10080514
Change view
Click to view available options
Type:
Epoxy Refillable Triode
Epoxy Triode Low Maintenance
For Use With (Equipment):
Orion A+ Series Meters
Orion Star Series Meters
Unit Size:
Each
Product Code. Type For Use With (Equipment) unitSize
10080514 Epoxy Refillable Triode Orion Star Series Meters Each
10691033 Epoxy Triode Low Maintenance Orion A+ Series Meters Each
11732857 Epoxy Triode Low Maintenance Orion Star Series Meters Each
10315173 Epoxy Refillable Triode Orion A+ Series Meters Each
4 options
Product Code. 10080514
Brand: Thermo Scientific™ 9180BNMD
Only null left
Add to basket
Add to basket
Accurately measure ORP and temperature together in just one electrode, with a built-in temperature sensor.
Thermo Scientific™ Orion™ Triode 3-in-1 pH/Automatic Temperature Compensation Electrodes come with a built-in ATC and are engineered with an epoxy body for ruggedness. The 9179BN and 9179BNMD are low-maintenance gel-filled electrodes, best suited for educational institutes where minimal maintenance is required, while the 9180BNMD is a refillable electrode best suited for general purpose samples.
Features:
  • Available in epoxy triode low maintenance and epoxy refillable triode construction
  • Built-in thermistor for Automatic Temperature Compensation (ATC)
  • Not designed for amperometric or constant current titrations, such as Karl Fischer analysis
TRUSTED_SUSTAINABILITY
Specifications
Description Epoxy Refillable Triode enance
Material Epoxy
For Use With (Equipment) Orion Star Series Meters
Type Epoxy Refillable Triode