Learn More
Thermo Scientific™ Orion™ Probe Maintenance Kits for Dissolved Oxygen Meters
Product Code. 10336112
Change view
Click to view available options
For Use With (Equipment):
Orion 080510MD, 081010MD, 08101 Probes
Orion 830A Series Probes
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
10336112 Orion 080510MD, 081010MD, 08101 Probes Each
10152962 Orion 830A Series Probes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10336112 Supplier Thermo Scientific™ Supplier No. 080113
  • ONLINE EXCLUSIVE PRICE
    1555.00 NOK
    1438.95 NOK / Pack of 1
    Save 116.05 NOK
    7% Off
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
Help ensure optimal performance of dissolved oxygen probes with these specialized probe maintenance kits.
Help ensure optimal performance of Orion polarographic dissolved oxygen probes with Thermo Scientific™ Orion™ Probe Maintenance Kits for Dissolved Oxygen Meters, which include electrolyte solution, polishing disk and two membrane caps.
Warranty: Out of box.
TRUSTED_SUSTAINABILITY
Specifications
Description For Orion 080510MD, 081010MD, 08101 Probes
For Use With (Equipment) Orion 080510MD, 081010MD, 08101 Probes
Type Probe Maintenance Kit
Includes 2 Membrane caps, electrolyte solution, and polishing disk
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.