Learn More
Thermo Scientific™ Orion™ Potassium Ion Selective Electrodes
Product Code. 10757352
Change view
Click to view available options
Connector Types:
BNC
BNC (Waterproof)
None
For Use With (Equipment):
Combination Electrode
Half-Cell Electrode
ISE Meter
Type:
Combination Electrode
Half Cell Electrode
Replacement Module
Unit Size:
Each
Product Code. Connector Types For Use With (Equipment) Type unitSize
10757352 BNC (Waterproof) ISE Meter Combination Electrode Each
10282871 None Half-Cell Electrode Replacement Module Each
10410643 BNC ISE Meter Half Cell Electrode Each
10390832 None Combination Electrode Replacement Module Each
4 options
Product Code. 10757352
Brand: Thermo Scientific™ 9719BNWP
Only null left
Add to basket
Add to basket
Measure potassium ions in aqueous solutions simply, accurately and economically.
Measure potassium ions in aqueous solutions simply, accurately and economically with Thermo Scientific™ Orion™ Potassium Electrodes.
  • Calibration Standard Catalog number: 0.1M KCl / 921906
  • Required ISA Catalog number: 931911
TRUSTED_SUSTAINABILITY
Specifications
Connector Types BNC (Waterproof)
Description Potassium Sure-Flow Combination Electrode with Waterproof BNC connector
Drift drift-free potentials
Junction Sure-Flow Junction
For Use With (Equipment) ISE Meter
Type Combination Electrode
Parameter Potassium