Learn More
Thermo Scientific™ Orion™ Potassium Ion Selective Electrodes
Product Code. 10410643
Change view
Click to view available options
Connector Types:
BNC
BNC (Waterproof)
None
For Use With (Equipment):
Combination Electrode
Half-Cell Electrode
ISE Meter
Type:
Combination Electrode
Half Cell Electrode
Replacement Module
Unit Size:
Each
Product Code. Connector Types For Use With (Equipment) Type unitSize
10410643 BNC ISE Meter Half Cell Electrode Each
10282871 None Half-Cell Electrode Replacement Module Each
10390832 None Combination Electrode Replacement Module Each
10757352 BNC (Waterproof) ISE Meter Combination Electrode Each
4 options
Product Code. 10410643
Brand: Thermo Scientific™ 9319BN
Only null left
Add to basket
Add to basket
Measure potassium ions in aqueous solutions simply, accurately and economically.
Measure potassium ions in aqueous solutions simply, accurately and economically with Thermo Scientific™ Orion™ Potassium Electrodes.
  • Calibration Standard Catalog number: 0.1M KCl / 921906
  • Required ISA Catalog number: 931911
TRUSTED_SUSTAINABILITY
Specifications
Connector Types BNC
Description Potassium Half Cell Electrode with BNC connector
Drift drift-free potentials
Junction Sure-Flow Junction
For Use With (Equipment) ISE Meter
Type Half Cell Electrode
Parameter Potassium