Learn More
Thermo Scientific™ Orion™ Potassium Ion Selective Electrodes
Description
Measure potassium ions in aqueous solutions simply, accurately and economically with Thermo Scientific™ Orion™ Potassium Electrodes.
- Calibration Standard Catalog number: 0.1M KCl / 921906
- Required ISA Catalog number: 931911
Specifications
Specifications
| Connector Types | None |
| Description | Replacement Module for Potassium Half-Cell Electrode 9319BN |
| Drift | drift-free potentials |
| Junction | Sure-Flow Junction |
| For Use With (Equipment) | Half-Cell Electrode |
| Type | Replacement Module |
| Parameter | Potassium |