Learn More
Thermo Scientific™ Orion™ Metallic Combination Electrodes
Perform accurate Karl Fischer titrations with these metallic combination electrodes.
Brand: Thermo Scientific™ 977900
Additional Details : Weight : 1.00000kg
Description
Efficient and reliable ORP/ redox measurement can be performed direct or with potentiometric titrations. 9678BNWP has a Sure-Flow junction that prevents clogging in dirty, viscous or wastewater samples. The epoxy body is suitable for rugged environments. 9778BNWP has a glass body that is chemical resistant that is excellent in samples with organic solvents.
Warranty: 12 months from date of purchase by Buyer or 18 months from date of shipment from Seller, whichever is earlier.Specifications
Model 977900; Double platinum KF glass electrode | |
Dual Platinum | |
Double Platinum KF Electrode, Glass | |
Orion Meters |
Double | |
0°C to 40°C | |
KF coulometric titrations | |
32°F to 104°F |