Learn More
Thermo Scientific™ Orion™ Ionplus™ Sure-Flow™ Lead Combination Ion Selective Electrode
Description
Measure lead ions in aqueous solutions quickly, simply, accurately and economically with Thermo Scientific™ Orion™ Lead Electrodes.
Features
Free-flowing Themo Scientific™ Sure-Flow™ junction assures stable, drift-free potentials. When conventional electrode junctions clog, the Sure-Flow junction can be opened and flushed clean simply by pressing the cap The lead electrode makes a sensitive endpoint detector for the titration of sulfate ions with lead perchlorate.
Solutions required:
Standard
948206 – 0.1 M lead standard, 475 mL bottle
948207 – 0.1 M sodium sulfate standard for sulfateanalysis with the lead ISE, 475 mL bottle
Electrode Fill Solution
900062 – Optimum Results B electrode filling solution, 5 x 60 mL bottles
Items included
900062 – Optimum Results B electrode filling solution, 60 mL 1 bottle
948201 – Polishing strip. Qty 2.
Warranty period
12 months from date of purchase
Specifications
Specifications
| Connector Types | BNC (Waterproof) |
| Description | With waterproof BNC connector |
| Drift | drift-free potentials |
| Junction | Sure-Flow Junction |
| For Use With (Equipment) | ISE Meter |
| Type | Lead Ion Selective Electrode, BNC waterproof connector |