Learn More
Thermo Scientific™ Orion™ Ionplus™ Sure-Flow™ Lead Combination Ion Selective Electrode
Product Code. 10191251
Change view
Click to view available options
Connector Types:
BNC
BNC (Waterproof)
Screw Cap
Unit Size:
Each
Product Code. Connector Types unitSize
10191251 BNC Each
10479102 Screw Cap Each
10587401 BNC (Waterproof) Each
3 options
Product Code. 10191251
Brand: Thermo Scientific™ 9482BN
Only null left
Add to basket
Add to basket
Measure lead ions in aqueous solutions quickly, simply, accurately and economically.
Measure lead ions in aqueous solutions quickly, simply, accurately and economically with Thermo Scientific™ Orion™ Lead Electrodes.
  • Free-flowing Themo Scientific™ Sure-Flow™ junction assures stable, drift-free potentials
  • When conventional electrode junctions clog, the Sure-Flow junction can be opened and flushed clean simply by pressing the cap
  • Lead Half-Cell Electrode used in conjunction with the Orion 900200 Double Junction Reference electrode offers the benefits of the Sure-Flow™ junction design
  • Calibration standards catalog number: 948206 - 0.1 M Pb(CIO4)2; 948207 - 0.1 M Na2SO4
  • Required ISA Catalog numbers: Consult instruction manual
TRUSTED_SUSTAINABILITY
Specifications
Connector Types BNC
Description With BNC connector
Drift drift-free potentials
Junction Sure-Flow Junction
For Use With (Equipment) ISE Meter
Type Lead Ion Selective Electrode, BNC connector