Learn More
Thermo Scientific™ Orion™ 9121APWP KNIpHE™ Replacement pH Electrode
Perform accurate determination of pH in meat matrices with this pH electrode.
Brand: Thermo Scientific™ 9121APWP
2471.88 NOK valid until 2024-04-30
Use promo code "21656" to get your promotional price.
Additional Details : CAS Number : 7447-40-7 Weight : 0.01000kg
Description
Perform accurate determination of pH in meat matrices with the Thermo Scientific™ Orion™ 9120APWP KNIpHE™ pH Electrode. It eliminates breakage with a stainless steel cutting blade surrounding a special combination pH electrode.
Perform accurate determination of pH in meat matrices with the Thermo Scientific™ Orion™ 9120APWP KNIpHE™ pH Electrode. It eliminates breakage with a stainless steel cutting blade surrounding a special combination pH electrode.
Specifications
Orion 9121APWP Double Junction Replacement KNIpHE Combination pH electrode | |
Ceramic | |
0°C to 60°C | |
3M KCl | |
Ag/AgCl Double Junction | |
32°F to 140°F |
BNC | |
0 to 14 | |
Combination pH Electrode, Replacement | |
Orion pH Meter | |
Store electrode in Orion pH electrode storage solution 910001 |