Learn More
Thermo Scientific™ Orion™ Gas-Sensing ISE membranes
Product Code. 10758971
Click to view available options
For Use With (Equipment):
Carbon Dioxide Electrode, 9502BNMP
HP Ammonia Electrode, 9512HPBNWP
HP Ammonia Electrode, 9512HPBNWP, 9512HP
Standard Ammonia Electrode, 9512BNWP, 9512
Unit Size:
Each
Pack of 20
Pack of 3
Product Code. 10758971
Brand: Thermo Scientific™ 951205
Only null left
Add to basket
Add to basket
Choose from loose or preassembled membranes for ease of use with Thermo Scientific™ Orion™ Gas-Sensing ISE membranes.
Choose from loose or preassembled membranes for ease of use with Thermo Scientific™ Orion™ Gas-Sensing ISE membranes.
Fresh membranes provide optimum performance with gas-sensing ion selective electrodes. Ammonia electrodes require different membranes if using standard electrode or high performance ammonia electrode.
TRUSTED_SUSTAINABILITY
Specifications
Description Gas-Sensing ISE membrane
For Use With (Equipment) Standard Ammonia Electrode, 9512BNWP, 9512
Type Gas-Sensing ISE Membrane
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis