Learn More
Thermo Scientific™ Orion™ Electrolyte Solution for Polarographic Dissolved Oxygen Probes
Product Code. 10037312
Change view
Click to view available options
Quantity:
60 mL Bottle
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10037312 60 mL Bottle Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10037312 Supplier Thermo Scientific™ Supplier No. 080514
  • ONLINE EXCLUSIVE PRICE
    223.00 NOK
    212.92 NOK / Pack of 1
    Save 10.08 NOK
    4% Off
 Restricted Item
Estimated Shipment: 11-11-2026
Maintain DO probes in optimal working condition with Thermo Scientific™ Orion™ Electrolyte Solution for Polarographic Dissolved Oxygen Probes.
Maintain DO probes in optimal working condition with Thermo Scientific™ Orion™ Electrolyte Solution for Polarographic Dissolved Oxygen Probes, available in a convenient 60mL bottle.
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 60 mL
Description Polargraphic Electrolyte Solution
For Use With (Equipment) Polarographic DO Probes
Type Electrolyte Solution
Quantity 60 mL Bottle