Learn More
Thermo Scientific™ Orion™ Ionplus™ Sure-Flow™ Cyanide Combination Ion Selective Electrode
Click to view available options
Connector Types:
BNC
BNC (Waterproof)
Screw Cap
Unit Size:
Each
Description
Measure cyanide ions in aqueous solutions quickly, simply and economically with Thermo Scientific™ Orion™ Cyanide Electrodes.
No separate reference electrode is needed with this electrode, making it convenient to use with small sample volumes.
- Calibration Standards Catalog number: Consult instruction manual
- Required ISA Catalog number: 951011
Specifications
Specifications
| Connector Types | Screw Cap |
| Description | With screw cap connector |
| Junction | Sure-Flow Junction |
| For Use With (Equipment) | ISE Meter |
| Type | Combination ISE |