Learn More
Thermo Scientific™ Orion™ Cupric Ion Selective Electrodes
Product Code. 10131211
Click to view available options
Connector Types:
BNC
BNC (Waterproof)
Screw Cap
Unit Size:
Each
Product Code. 10131211
Brand: Thermo Scientific™ 9429SC
Only null left
Add to basket
Add to basket
Measure cupric ions in aqueous solutions quickly, simply, accurately and economically.
Measure cupric ions in aqueous solutions quickly, simply, accurately and economically with the Thermo Scientific™ Orion™ Cupric Electrodes.
No separate reference electrode is needed with this electrode, making it convenient to use with small sample volumes.
  • Calibration Standards Catalog number: 0.1 M Cu(NO3)2/942906
  • Required ISA Catalog number: 940011
TRUSTED_SUSTAINABILITY
Specifications
Connector Types Screw Cap
Description With screw cap connector
Junction Sure-Flow Junction
For Use With (Equipment) ISE Meter
Type Combination ISE