Learn More
Thermo Scientific™ Orion™ Cupric Ion Selective Electrodes
Product Code. 10049150
Click to view available options
Connector Types:
BNC
BNC (Waterproof)
Screw Cap
Unit Size:
Each
Product Code. 10049150
Brand: Thermo Scientific™ 9629BNWP
Only null left
Add to basket
Add to basket
Measure cupric ions in aqueous solutions quickly, simply, accurately and economically.
Measure cupric ions in aqueous solutions quickly, simply, accurately and economically with the Thermo Scientific™ Orion™ Cupric Electrodes.
No separate reference electrode is needed with this electrode, making it convenient to use with small sample volumes.
  • Calibration Standards Catalog number: 0.1 M Cu(NO3)2/942906
  • Required ISA Catalog number: 940011
TRUSTED_SUSTAINABILITY
Specifications
Connector Types BNC (Waterproof)
Description With waterproof BNC connector BNC-to-USS adapter is required with this product as a onetime purchase. Part Number: 13-641-890
Junction Sure-Flow Junction
For Use With (Equipment) ISE Meter
Type Combination ISE