Learn More
Thermo Scientific™ Orion™ Single Junction Thin Stem Glass Bodied Combination pH Electrode
Product Code. 10738591
Click to view available options
Connector Types:
BNC (Waterproof)
Screw Cap
Unit Size:
Each
Product Code. 10738591
Brand: Thermo Scientific™ 9103SC
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Top-performance, semi-micro pH electrodes to measure samples as small as 0.2 mL
Thermo Scientific™ Orion™ Silver Chloride Semi-Micro Electrodes provide reliable and accurate measurements of routine samples. They are available in chemically resistant glass bodies and best suited for measuring general samples in test tubes and small containers.
Includes:
  • 60ml bottle of Orion filling solution, 900011
TRUSTED_SUSTAINABILITY
Specifications
Description With screw cap connector
Connector Types Screw Cap
Junction Ceramic
Material Glass Body
pH Range 0 to 14
Temperature (Metric) Operating 0°C to 90°C
Type Combination pH Electrode
Filling Solution Type 4M KCl with Ag/AgCl
Internal Reference Ag/AgCl
Precision 0.02
Temperature (English) Operating 32°F to 194°F
Diameter (Metric) Cap 16 mm