Learn More
Thermo Scientific™ Orion™ ROSS™ Semi-Micro Spear Tip Glass Bodied Combination pH Electrode
Product Code. 10304721
Click to view available options
Connector Types:
BNC
Unit Size:
Each
Product Code. 10304721
Brand: Thermo Scientific™ 8163BNWP
Only null left
Add to basket
Add to basket
Pierce soft and semi-solid samples like cheese, meat, or fruit with a refillable spear tip pH electrode.
Thermo Scientific™ Orion™ ROSS Spear Tip Electrodes provide optimum accuracy and stability while having the additional advantages of a double junction design. Best suited for piercing solid/semi-solid samples and measuring small volume samples.
Features
  • For pH measurement in semi-solid food material, such as cheese, meat, fruit, bread and other similar samples
  • Works in extreme case of samples varying from one another by 50°C or more
  • Drift is less than 0.002 pH per day; restandardization is minimized
Includes:
  • 60ml bottle of ROSS electrode fill solution, 810007
TRUSTED_SUSTAINABILITY
Specifications
Description With BNC waterproof connector
Connector Types BNC
Junction Ceramic
Material Glass Body
pH Range 0 to 14
Temperature (Metric) Operating 0°C to 100°C
Type Combination Spear Tip pH Electrode, BNC waterproof connector
Drift <0.002pH/day
Filling Solution Type 3M KCl
For Use With (Equipment) Standard pH Meter, BNC connector
Includes 60mL bottle of ROSS electrode fill solution
Internal Reference ROSS
Precision 0.01
Sample Size 100 μL
Temperature (English) Operating 32°F to 212°F
Tip Type Spear
Length (Metric) Tip 20 mm