missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ ORC4L (Human) Recombinant Protein
Human ORC4L full-length ORF ( AAH05388, 1 a.a. - 248 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00005000-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MSSRKSKSNSLIHTECLSQVQRILRERFCRQSPHSNLFGVQVQYKHLSELLKRTALHGESNSVLIIGPRGSGKTMLISHALKELMEIEEVSENVLQVHLNGLLQINDKIALKEITRQLNLENVVGDKVFGSFAENLSFLLEALKKGDRTSSCPVIFILDEFDLFAHHKNQTLLYNLFDISQSAQTPIAVIGLTCRLDILELLEKRVKSRFSHRQIHLMNSFGFPQYVKIFKEQLSLPAEFPDKVFAEK
Specifications
AAH05388 | |
origin recognition complex, subunit 4-like (yeast) | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ORC4L | |
GST |
5000 | |
Wheat germ expression system | |
25 μg | |
ORC4, ORC4P | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |