missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oligomycin, >98%, For TLC analysis, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
Specifications
Specifications
| Type | Oligomycin |
| Physical Form | Powder |
| Color | White |
| Quantity | 10 mg |
| Formula Weight | ≈789.3 |
| Percent Purity | >98% |
| Chemical Name or Material | Oligomycin, For TLC analysis |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction