CAS RN 1404-19-9
CAS RN 1404-19-9
IUPAC Name:
(4E,18E,20Z)-22-ethyl-7,11,14,15-tetrahydroxy-6'-(2-hydroxypropyl)-5',6,8,10,12,14,16,28,29-nonamethyl-2,26-dioxaspiro[bicyclo[23.3.1]nonacosane-27,2'-oxane]-4,18,20-triene-3,9,13-trione
Synonyms:
oligomycin
Molecular Weight (g/mol):
791.08
Molecular Formula:
C45H74O11
InChi Key:
MNULEGDCPYONBU-ZUSSGZTJNA-N
SMILES:
CCC1CCC2OC3(CCC(C)C(CC(C)O)O3)C(C)C(OC(=O)\C=C\C(C)C(O)C(C)C(=O)C(C)C(O)C(C)C(=O)C(C)(O)C(O)C(C)C\C=C\C=C/1)C2C
1
–
3
of
3
results
2
Oligomycin, >98%, For TLC analysis, MP Biomedicals™
CAS: 1404-19-9 Molecular Formula: C45H74O11 Molecular Weight (g/mol): 791.08 MDL Number: MFCD00065705,MFCD01779388 InChI Key: MNULEGDCPYONBU-ZUSSGZTJNA-N Synonym: Olivomycin,Aburamycin PubChem CID: 78358496 IUPAC Name: (4E,18E,20Z)-22-ethyl-7,11,14,15-tetrahydroxy-6'-(2-hydroxypropyl)-5',6,8,10,12,14,16,28,29-nonamethyl-2,26-dioxaspiro[bicyclo[23.3.1]nonacosane-27,2'-oxane]-4,18,20-triene-3,9,13-trione SMILES: CCC1CCC2OC3(CCC(C)C(CC(C)O)O3)C(C)C(OC(=O)\C=C\C(C)C(O)C(C)C(=O)C(C)C(O)C(C)C(=O)C(C)(O)C(O)C(C)C\C=C\C=C/1)C2C