Learn More
Ohaus™ OIML E1 Weight Set, NVLAP Certificate
Supplied with a NVLAP Accredited Certificate and Adjusted to Meet or Exceed OIML E1 Tolerances
18655.00 NOK - 154280.00 NOK
Specifications
Certifications/Compliance | NVLAP Accredited Certificate |
---|---|
Includes | Protective Case |
Product Code | Brand | Mass | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Mass | Price | Quantity & Availability | |||||
16612282
|
Ohaus™
30390926 |
500 mg to 1 mg |
18655.00 NOK
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
|||||
16682272
|
Ohaus™
30390922 |
100 g to 1 mg |
43405.00 NOK
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
|||||
16612202
|
Ohaus™
30390040 |
2 x 200 g to 1 mg |
56300.00 NOK
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
|||||
16662272
|
Ohaus™
30390918 |
500 g to 1 mg |
60445.00 NOK
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
|||||
16652272
|
Ohaus™
30390917 |
500 g to 1 g |
47780.00 NOK
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
|||||
16642272
|
Ohaus™
30390916 |
1 kg to 1 mg |
71615.00 NOK
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
|||||
16632272
|
Ohaus™
30390915 |
1 kg to 1 g |
54000.00 NOK
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
|||||
16692192
|
Ohaus™
30390038 |
2 x 2 kg to 1 mg |
100630.00 NOK
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
|||||
16682192
|
Ohaus™
30390037 |
2 x 2 kg to 1 g |
82895.00 NOK
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
|||||
16622272
|
Ohaus™
30390912 |
5 kg to 1 mg |
127800.00 NOK
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
|||||
16612272
|
Ohaus™
30390911 |
5 kg to 1 g |
107880.00 NOK
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
|||||
16602272
|
Ohaus™
30390909 |
10 kg to 1 mg |
154280.00 NOK
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
|||||
16692262
|
Ohaus™
30390908 |
10 kg to 1 g |
140465.00 NOK
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
|||||
16692272
|
Ohaus™
30390923 |
50 g to 1 g |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16602202
|
Ohaus™
30390039 |
2 x 200 g to 1 g |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16602282
|
Ohaus™
30390924 |
50 g to 1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16672272
|
Ohaus™
30390921 |
100 g to 1 g |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Descripción
- OIML Class E1 offers high accuracy with tight tolerances and guaranteed uncertainty 1/3 of the tolerance
- Weight surfaces meet or exceed OIML R 111 and have a highly polished mirror-like finish
- Weights in masses ranging from 1 gram to 20 kilograms are made of Troemner Alloy 8 Stainless Steel which possesses closely controlled density (8.03 g/cm3), extremely low magnetic properties, good stability, and resistance to corrosion
- Weights are supplied in high quality cases to ensure weights are held securely, kept clean, and dust free
- NVLAP Calibration Certificate is accredited to ISO 17025 which is recognizable through ILAC
Especificaciones
NVLAP Accredited Certificate | |
Protective Case |