Learn More
Ohaus™ OIML E1 Weight Set
Adjusted to Meet or Exceed OIML E1 Tolerances
Brand: Ohaus™ 30518003
Description
- OIML Class E1 offers high accuracy with tight tolerances and guaranteed uncertainty 1/3 of the tolerance
- Weight surfaces meet or exceed OIML R 111 and have a highly polished mirror-like finish
- Weights in masses ranging from 1 gram to 20 kilograms are made of Troemner Alloy 8 Stainless Steel which possesses closely controlled density (8.03 g/cm3), extremely low magnetic properties, good stability, and resistance to corrosion
- Weights are supplied in high quality cases to ensure weights are held securely, kept clean, and dust free, protecting your investment
Specifications
No Certificate | |
10 kg to 1 g | |
17/Pk. |
Troemner Alloy 8 | |
Protective Case |