Learn More
Ohaus™ OIML E1 Weight Set
Adjusted to Meet or Exceed OIML E1 Tolerances
Brand:  Ohaus™ 30517990
Additional Details : Weight : 0.60000kg
 View more versions of this product
Product Code. 16612332
  • 33045.00 NOK / Each
Estimated Shipment: 07-06-2024
to see stock.
Add to basket
Description
Description
  • OIML Class E1 offers high accuracy with tight tolerances and guaranteed uncertainty 1/3 of the tolerance
  • Weight surfaces meet or exceed OIML R 111 and have a highly polished mirror-like finish
  • Weights in masses ranging from 1 gram to 20 kilograms are made of Troemner Alloy 8 Stainless Steel which possesses closely controlled density (8.03 g/cm3), extremely low magnetic properties, good stability, and resistance to corrosion
  • Weights are supplied in high quality cases to ensure weights are held securely, kept clean, and dust free, protecting your investment
Specifications
Specifications
No Certificate
2 x 200 g to 1 mg
23/Pk.
Troemner Alloy 8, Stainless Steel, Aluminum
Protective Case