Learn More
O-Terphenyl, TRC
Product Code. 30137355
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137355 5 g 5g
30135110 25 g 25g
18044050 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30137355 Supplier Toronto Research Chemicals Supplier No. TRCT1166855G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 84-15-1
Molecular Formula C18 H14
Molecular Weight (g/mol) 230.3
Synonym 1,1':2',1' '-Terphenyl, o-Terphenyl (8CI), 1,1'-Biphenyl, 2-phenyl-, 1,2-Diphenylbenzene, NSC 6809
IUPAC Name 1,2-diphenylbenzene
SMILES c1ccc(cc1)c2ccccc2c3ccccc3
Specifications
Quantity 5 g
InChI Formula InChI=1S/C18H14/c1-3-9-15(10-4-1)17-13-7-8-14-18(17)16-11-5-2-6-12-16/h1-14H
Formula Weight 230.1096
Chemical Name or Material o-Terphenyl