CAS RN 84-15-1
IUPAC Name:
2-phenyl-1,1'-biphenyl
Synonyms:
O-terphenyl
Molecular Weight (g/mol):
230.31
Molecular Formula:
C18H14
InChi Key:
OIAQMFOKAXHPNH-UHFFFAOYSA-N
SMILES:
C1=CC=C(C=C1)C1=CC=CC=C1C1=CC=CC=C1
1
–
5
of
5
results
1
O-Terphenyl, TRC
CAS: 84-15-1 Molecular Formula: C18 H14 Molecular Weight (g/mol): 230.3 Synonym: 1,1':2',1' '-Terphenyl,o-Terphenyl (8CI),1,1'-Biphenyl, 2-phenyl-,1,2-Diphenylbenzene,NSC 6809 IUPAC Name: 1,2-diphenylbenzene SMILES: c1ccc(cc1)c2ccccc2c3ccccc3
2
o-Terphenyl, Sigma-Aldrich™
CAS: 84-15-1
3
o-Terphenyl 10000 μg/mL in Dichloromethane, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality