Learn More
Cole-Parmer™ Nylon Manifold, 4 outlets
Expand your flow and line capacity
725.00 NOK - 1080.00 NOK
Specifications
Product Type | Nylon Manifold, 4 Outlets |
---|---|
Material | Nylon |
Min. Operating Temperature (English) | -60°F |
Min. Operating Temperature (Metric) | -54°C |
Maximum Pressure | 200 psi |
Product Code | Brand | Tubing Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Tubing Size | Price | Quantity & Availability | |||||
11795218
|
Cole-Parmer™
31522-39 |
3/8 in. |
725.00 NOK
Each |
Estimated Shipment: 08-07-2024
Log in to see stock. |
|||||
11705228
|
Cole-Parmer™
31522-41 |
1/2 in. |
1080.00 NOK
Each |
Estimated Shipment: 04-07-2024
Log in to see stock. |
|||||
Description
- Constructed of nylon for economic general applications.
- Offers good chemical compatibility.
- Available in a variety of configurations to suit a wide range of applications.
Specifications
Nylon Manifold, 4 Outlets | |
-60°F | |
200 psi | |
82°C |
Nylon | |
-54°C | |
180°F |