Learn More
Cole-Parmer™ Nylon Manifold, 4 outlets
Expand your flow and line capacity
Brand: Cole-Parmer™ 31522-41
Additional Details : Weight : 0.80000kg
Description
- Constructed of nylon for economic general applications.
- Offers good chemical compatibility.
- Available in a variety of configurations to suit a wide range of applications.
Specifications
Nylon Manifold, 4 Outlets | |
4.75 in. | |
-60°F | |
1/2 in. | |
3/8 in. NPT(F) | |
1.5 in. | |
1.5 in. | |
180°F |
Nylon | |
12.07 cm | |
-54°C | |
1/2 in. NPT(F) | |
3.81 cm | |
3.81 cm | |
200 psi | |
82°C |