Learn More
Biodesign™ Nylon CellMicroSieves Rolls
Product Code. 12974267
Click to view available options
Pore Size:
10 μm
100um
15 μm
25 μm
50 μm
8 μm
Unit Size:
Each
Product Code. 12974267
Brand: Biodesign™ N15R
Only null left
Add to basket
Add to basket
Autoclavable and can be repeatedly washed and reused
Unlike traditional filters, CellMicroSieves™ are a precision woven matrix of pure nylon filaments that is available with pores ranging from 5 μm to 200 μm. CellMicroSieves™ do not wet and contains no dyes, detergents or extractables. Extremely low protein affinity means that cells and proteins will not bind to CellMicroSieves™. They have a wide variety of distinctive uses a specific sized porous screens in the laboratory.
AccessoriesPolypropylene sieve holders (2 piece) holders with adjustable metal tightener or filtration units, type A borosilicate glass with Stainless Steel filter support and aluminum clamp or type B complete polysulfone filtration unit with polypropylene filter support are available. Both filtration units have a 300 mL capacity are autoclavable and reusable.Use sheets and rolls with sieve holders and circles with filtration units to hold securely when processing samples.
  • A pure nylon product that is biologically inert, resistant to most acidic or basic conditions and many solvents
  • Autoclavable and can be repeatedly washed and reused
  • Rapid collection of tissue culture cells without centrifugation
  • Isolation of specific cell types from a dissociated tissue sample, allowing only a single type of cell to pass through
  • Separation of different sized cells from a mixture of cell types
  • Concentration and isolation of cells from a bioreactor
  • Prefilter to prevent clogging of final filter
CellMicroSieves™ allow the rapid gentle isolation of cells, organelles or cellular debris.
Specifications
Quantity Each
Pore Size 15 μm
Type Cell Micro Sieve
Material Nylon
For Use With (Application) Isolation of Specific Cell Types From a Dissociated Tissue Sample
Format Roll