Learn More
Biodesign™ Nylon CellMicroSieves Circles
Product Code. 12914287
Change view
Click to view available options
Pore Size:
5 μm
8 μm
10 μm
15 μm
35 μm
Unit Size:
Pack of 10
Product Code. Pore Size unitSize
12914287 5 μm Pack of 10
12994277 10 μm Pack of 10
12984277 15 μm Pack of 10
12954277 35 μm Pack of 10
12904287 8 μm Pack of 10
5 options
Product Code. 12914287
Brand: Biodesign™ N5C
Only null left
Add to basket
Add to basket
Autoclavable and can be repeatedly washed and reused
Unlike traditional filters, CellMicroSieves™ are a precision woven matrix of pure nylon filaments that is available with pores ranging from 5 μm to 200 μm. CellMicroSieves™ do not wet and contains no dyes, detergents or extractables. Extremely low protein affinity means that cells and proteins will not bind to CellMicroSieves™. They have a wide variety of distinctive uses a specific sized porous screens in the laboratory.
AccessoriesPolypropylene sieve holders (2 piece) holders with adjustable metal tightener or filtration units, type A borosilicate glass with Stainless Steel filter support and aluminum clamp or type B complete polysulfone filtration unit with polypropylene filter support are available. Both filtration units have a 300 mL capacity are autoclavable and reusable.Use sheets and rolls with sieve holders and circles with filtration units to hold securely when processing samples.
  • A pure nylon product that is biologically inert, resistant to most acidic or basic conditions and many solvents
  • Autoclavable and can be repeatedly washed and reused
  • Rapid collection of tissue culture cells without centrifugation
  • Isolation of specific cell types from a dissociated tissue sample, allowing only a single type of cell to pass through
  • Separation of different sized cells from a mixture of cell types
  • Concentration and isolation of cells from a bioreactor
  • Prefilter to prevent clogging of final filter
CellMicroSieves™ allow the rapid gentle isolation of cells, organelles or cellular debris.
Specifications
Quantity 10/Pk.
Pore Size 5 μm
Type Cell Micro Sieve
Material Nylon
For Use With (Application) Isolation of Specific Cell Types From a Dissociated Tissue Sample
Format Circle