Learn More
Novus Biologicals™ NXPH4 293T Cell Transient Overexpression Lysate
Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
Specifications
Specifications
| Quantity | 0.1 mL |
| Content And Storage | Store at -80C. Avoid freeze-thaw cycles. |
| Research Category | Neuroscience |
| Target | NXPH4 |
| Synonym | neurexophilin 4, NPH4neurexophilin-4 |
| Gene Symbol | NXPH4 |