missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nucleoside phosphorylase Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18009-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This gene is a member of the DEAH box polypeptide family. The encoded protein contains the DEAH (Asp-Glu-Ala-His) motif which is characteristic of all DEAH box proteins, and is thought to function as an ATP-dependent RNA helicase that regulates the release of spliced mRNAs from spliceosomes prior to their export from the nucleus. This protein may be required for the replication of human immunodeficiency virus type 1 (HIV-1). Alternative splicing results in multiple transcript variants.Specifications
Nucleoside phosphorylase | |
Polyclonal | |
Unconjugated | |
PNP | |
Inosine phosphorylase, NP, nucleoside phosphorylase, PNP, PRO1837, PUNP | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
18950, 290029, 4860 | |
-20°C | |
Liquid |
Western Blot | |
0.46 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P00491, P23492, P85973 | |
Pnp | |
Nucleoside phosphorylase Fusion Protein Ag12507 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |