missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nucleoside phosphorylase Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | Nucleoside phosphorylase |
---|---|
Concentration | 0.46 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16898034
|
Proteintech
18009-1-AP-20UL |
20 μL |
1611.84 NOK
20µL |
Estimated Shipment: 06-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16888034
|
Proteintech
18009-1-AP-150UL |
150 μL |
4593.76 NOK
150µL |
Estimated Shipment: 06-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene is a member of the DEAH box polypeptide family. The encoded protein contains the DEAH (Asp-Glu-Ala-His) motif which is characteristic of all DEAH box proteins, and is thought to function as an ATP-dependent RNA helicase that regulates the release of spliced mRNAs from spliceosomes prior to their export from the nucleus. This protein may be required for the replication of human immunodeficiency virus type 1 (HIV-1). Alternative splicing results in multiple transcript variants.Specifications
Nucleoside phosphorylase | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
P00491, P23492, P85973 | |
18950, 290029, 4860 | |
Nucleoside phosphorylase Fusion Protein Ag12507 | |
Primary | |
-20°C | |
PNP |
0.46 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Inosine phosphorylase, NP, nucleoside phosphorylase, PNP, PRO1837, PUNP | |
Pnp | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |