Learn More
Macherey-Nagel™ NucleoBond™ AXG 20, Mini columns for high integrity DNA
Product Code. 12748472
Change view
Click to view available options
Yield:
20 μg
Unit Size:
Pack of 20
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Yield Unit Size
12748472 20 μg Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12748472 Supplier Macherey-Nagel™ Supplier No. 740544
  • ONLINE EXCLUSIVE PRICE
    1410.00 NOK
    1259.89 NOK / Pack of 20
    Save 150.11 NOK
    10% Off
In Stock
Anion exchange chromatography for purification of DNA from bacteria, yeast, and tissue
Genomic DNA quality is extraordinarily high. Samples are first lysed under special conditions for removal of erythrocytes. After incubation with proteinase K, the sample is diluted with buffer N2 before loading onto the NucleoBond™ column. After elution with high-salt buffer the genomic DNA is precipitated with isopropanol. The DNA purified by this method is suitable for all kinds of subsequent reactions. Kit components: NucleoBond™ CB 100 kit - NucleoBond AXG 100 columns, buffers, proteinase K. NucleoBond™ CB 500 kit - NucleoBond™ AXG 500 columns, buffers, proteinase K.
  • Anion exchange technology
  • Format: gravity flow columns
  • Sample material: whole blood, buffy coat, cultured cells (up to 20 mL blood)
  • Fragment size: 500bp to 300kb
  • Yield: 100μg or 500μg depending on column used
  • Binding capacity: 100μg or 500μg depending on column used
  • Preparation time: 4 to 5hrs For purification of up to 500μg genomic DNA from whole blood.
Specifications
Final Product Type NucleoBond AXG 20 Columns
For Use With (Application) Genomic DNA from bacteria, yeast*, tissue
Product Type NucleoBond AXG 20 Columns
Quantity 1 Pack
Yield 20 μg