missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human NT-3 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 4 publications
1930.00 NOK - 18530.00 NOK
Specifications
| Antigen | NT-3 |
|---|---|
| Clone | 41512 |
| Dilution | Neutralization 0.1-0.5 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | Neutralization, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18793318
![]() |
R&D Systems
MAB267-SP |
25 μg |
1930.00 NOK
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18735669
![]() |
R&D Systems
MAB267-100 |
100 μg |
4680.00 NOK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18752709
![]() |
R&D Systems
MAB267-500 |
500 μg |
18530.00 NOK
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NT-3 Monoclonal specifically detects NT-3 in Human samples. It is validated for Neutralization, ELISA Capture (Matched Antibody Pair).Specifications
| NT-3 | |
| Neutralization 0.1-0.5 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| HDNF, MGC129711, Nerve growth factor 2, Neurotrophic factor, neurotrophin 3, neurotrophin-3, NGF2, NGF-2, NT3, NT-3, NTF3 | |
| NTF3 | |
| IgG1 | |
| Protein A or G purified from ascites | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 41512 | |
| Neutralization, ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| P20783 | |
| 4908 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human NT-3 Tyr139-Thr257 Accession # P20783 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human NT-3 in ELISAs. In sandwich immunoassays, no cross-reactivity with recombinant human (rh) BDNF, rhCNTF, rhGDNF, rh beta-NGF, and rhNT-4 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title