missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human NT-3 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 4 publications
Brand: R&D Systems MAB267-SP
This item is not returnable.
View return policy
Description
NT-3 Monoclonal specifically detects NT-3 in Human samples. It is validated for Neutralization, ELISA Capture (Matched Antibody Pair).
Specifications
| NT-3 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| HDNF, MGC129711, Nerve growth factor 2, Neurotrophic factor, neurotrophin 3, neurotrophin-3, NGF2, NGF-2, NT3, NT-3, NTF3 | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 4908 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Neutralization, ELISA | |
| 41512 | |
| Neutralization 0.1-0.5 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| P20783 | |
| NTF3 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human NT-3 Tyr139-Thr257 Accession # P20783 | |
| 25 μg | |
| Primary | |
| Detects human NT-3 in ELISAs. In sandwich immunoassays, no cross-reactivity with recombinant human (rh) BDNF, rhCNTF, rhGDNF, rh beta-NGF, and rhNT-4 is observed. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction