Learn More
NR3C2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 21854-1-AP-150UL
 View more versions of this product
Product Code. 16861405
  • 4836.10 NOK / 150µL
Description
This antibody is a rabbit polyclonal antibody, specifically reacts with the 107 kDa MCR protein.
Steroid receptors are ligand-dependent, intracellular proteins that stimulate transcription of specific genes by binding to specific DNA sequences following activation by the appropriate hormone. Mineralocorticoids are a family of steroids, secreted by the adrenal cortex, necessary for the regulation of a number of metabolic processes including electrolyte regulation. These compounds exert their effect through their interaction with the mineralocorticoid receptor (MR) and that complex′s subsequent association with DNA. Given the function of mineralocorticoids, it is not surprising to find that the kidney is a primary target organ for mineralocorticoids and that this organ has been shown to contain MR. The corresponding gene for the mineralocorticoid receptor is NR3C2.
Specifications
Specifications
NR3C2
Polyclonal
Unconjugated
NR3C2
MCR, Mineralocorticoid receptor, MLR, MR, NR3C2
Rabbit
Antigen Affinity Chromatography
RUO
110784, 25672, 4306
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.53 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P08235, P22199, Q8VII8
NR3C2
NR3C2 Fusion Protein Ag16410
150 μL
Primary
Human, Mouse, Rat