Learn More
NR2C2 Mouse anti-Human, Unconjugated, Clone: 2E4C10, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67489-1-IG-150UL
This item is not returnable.
View return policy
Description
The NR2C2 antibody from Proteintech is a mouse monoclonal antibody generated with peptide of human NR2C2. This antibody recognizes human, mouse, rat antigen. The NR2C2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| NR2C2 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse |
| Western Blot, ELISA | |
| 2E4C10 | |
| Unconjugated | |
| 150 μL | |
| 7182 | |
| IgG1 |