Learn More
NR2C2 Mouse anti-Human, Unconjugated, Clone: 2E4C10, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67489-1-IG-150UL
 View more versions of this product
Product Code. 16850146
  • 4836.10 NOK / 150µL
Description
The NR2C2 antibody from Proteintech is a mouse monoclonal antibody generated with peptide of human NR2C2. This antibody recognizes human, mouse, rat antigen. The NR2C2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
NR2C2
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, ELISA
2E4C10
Unconjugated
150 μL
7182
IgG1