missing translation for 'onlineSavingsMsg'
Learn More

NR2C2 Mouse anti-Human, Unconjugated, Clone: 2E4C10, Proteintech

Product Code. p-7223771 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16860146 20 μL 20µL
16850146 150 μL 150µL
2 options
This item is not returnable. View return policy

Product Code. 16860146

Brand: Proteintech 674891IG20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The NR2C2 antibody from Proteintech is a mouse monoclonal antibody generated with peptide of human NR2C2. This antibody recognizes human, mouse, rat antigen. The NR2C2 antibody has been validated for the following applications: WB, ELISA analysis.

Specifications

Antigen NR2C2
Applications Western Blot, ELISA
Classification Monoclonal
Clone 2E4C10
Concentration 1000 μg/mL
Conjugate Unconjugated
Host Species Mouse
Quantity 20 μL
Primary or Secondary Primary
Gene ID (Entrez) 7182
Target Species Human, Rat, Mouse
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.