Learn More
Invitrogen™ Mouse Tau (Phospho) [pS199] ELISA Kit
Product Code. 10272883
Click to view available options
Quantity:
96 Tests
Unit Size:
Each
Product Code. 10272883
Brand: Invitrogen™ KMB7041
ELISA
The Mouse Tau (Phospho) [pS199] ELISA quantitates Ms Tau (Phospho) [pS199] in mouse cell lysates, cell culture medium and brand homogenates. The assay will exclusively recognize both natural and recombinant Ms Tau (Phospho) [pS199]. Principle of the method The Ms Tau (Phospho) [pS199] solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, binding to the target on a different epitope from the capture antibody. A conjugated enzyme has been incorporated into the assay. After incubation and washing steps to rid the microplate of unbound substances, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Tau is a neuronal microtubule-associated protein found predominantly on axons. The function of Tau is to promote tubulin polymerization and stabilize microtubules. The C-terminus binds axonal microtubules while the N- terminus binds neural plasma membrane components, suggesting that tau functions as a linker protein between both. Axonal polarity is predetermined by TAU/MAPT localization (in the neuronal cell) in the domain of the cell body defined by the centrosome. The short isoforms allow plasticity of the cytoskeleton while the longer isoforms may preferentially play a role in its stabilization. In its hyper-phosphorylated form, Tau is the major component of paired helical filaments (PHF), the building block of neurofibrillary lesions in Alzheimer's diseases (AD) brain. Hyper-phosphorylation impairs the microtubule binding function of Tau, resulting in the destabilization of microtubules in AD brains, ultimately leading to the degeneration of the affected neurons. Numerous serine/threonine kinases phosphorylate Tau, including GSK-3beta, protein kinase A (PKA), cyclin-dependent kinase 5 (cdk5) and casein kinase II. Hyper-phosphorylated Tau is found in neurofibrillary lesions in a range and other central nervous system disorders such as Pick's disease, frontotemporal dementia, cortico-basal degeneration and progressive supranuclear palsy.
TRUSTED_SUSTAINABILITY
Specifications
Accession Number P10637
Assay Range 313 to 20,000 pg/mL
Assay Sensitivity <58.5 pg/mL
Concentration 15.6 to 1000 pg/mL
Conjugate HRP
Product Type ELISA
Sample Type Brain Homogenate, Cell Lysates, Supernatant
Target Species Mouse
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 17762
Gene Symbol AI413597,AW045860,Mtapt,Tau
Incubation Time 4 hr.
Interassay CV 3.8%
Intraassay CV 5.6
Kit Contents Pre-coated 96 well plate, Standard, Standard Dilution Buffer, Biotinylated Detection Antibody, Streptavidin-HRP, HRP Diluent, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Marker Gene Tau
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Brain Homogenate 10 μL, Cell Lysate 5 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target Tau
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs