Learn More
Invitrogen™ Novex™ Leukotriene B4 Human ELISA Kit
Product Code. 10627035
Click to view available options
Quantity:
96 Assays
Unit Size:
96 tests
Product Code. 10627035
Brand: Invitrogen™ KHL1741
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Includes
Pre-coated plate, standard, antibody, AP tracer, wash buffer, tris buffer, DEA buffer, pNPP tablets, and plate cover.
Competitive immunoassay to quantify LTB4 in biological samples
LTB4 is a potent inflammatory mediator that works by binding BLT1, on leukocytes.
Cell Analysis, Immunoassays, Ready-To-Use Immunoassay
Order Info
Shipping Condition: Room Temperature
TRUSTED_SUSTAINABILITY
Specifications
Accession Number NM_001143919.2, NM_181657.3, NP_001137391.1, NP_858043.1
Sample Type Plasma, Serum
For Use With (Application) Colorimetric Detection
For Use With (Equipment) Microplate Reader
Gene ID (Entrez) 1241
Incubation Time 3 hr.
Kit Contents Antibody, AP tracer, DEA buffer, Plate covers, pNPP tablets, Pre-coated plate, Standard, Tris buffer, Wash buffer
Marker Gene Leukotriene B4
Quantity 96 Assays
Target leukotriene B4 receptor