Learn More
Fisherbrand™ Notched Glass Plates with Bonded Spacers
Product Code. 11807643 Shop All Fisherbrand Products
Change view
Click to view available options
Size:
0.5 mm Bonded Spacers
0.75 mm Bonded Spacers
1.5 mm Bonded Spacers
2 mm Bonded Spacers
Unit Size:
Pack of 2
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
11807643 0.75 mm Bonded Spacers Pack of 2
11897633 0.5 mm Bonded Spacers Pack of 2
11827643 1.5 mm Bonded Spacers Pack of 2
11837643 2 mm Bonded Spacers Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11807643
Only null left
Add to basket
Add to basket
Accessory for use with OmniPAGE mini vertical Electrophoresis systems.
  • Accessory for use with OmniPAGE mini vertical Electrophoresis systems
Specifications
Type Notched Glass Plate
Gel Dimensions 100 x 100 mm
Size 0.75 mm Bonded Spacers