Learn More
Fisherbrand™ Notched Glass Plates
Product Code. p-8009027 Shop All Fisherbrand Products
Change view
Click to view available options
Thickness (Metric):
1 mm bonded spacers
2 mm
4 mm
Gel Dimensions:
10 x 10 cm
20 x 20 cm
Unit Size:
Pack of 2
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Thickness (Metric) Gel Dimensions unitSize
11887633 2 mm 10 x 10 cm Pack of 2
11817643 1 mm bonded spacers 10 x 10 cm Pack of 2
11854532 4 mm 20 x 20 cm Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11887633 Supplier Fisherbrand™ Supplier No. VS10NG
Only null left
Add to basket
Add to basket
Accessory for Fisherbrand OmniPAGE Vertical Electrophoresis Systems.
  • Accessory for Fisherbrand OmniPAGE Vertical Electrophoresis Systems
Specifications
For Use With (Equipment) Fisherbrand OmniPAGE Mini Vertical Electrophoresis System
Type Notched Glass Plate
Gel Dimensions 10 x 10 cm
Quantity 2/Pk.
Thickness (Metric) 2 mm