Learn More
Novus Biologicals™ NKX3-2 Overexpression Lysate
Product Code. 18790374 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18790374 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18790374 Supplier Novus Biologicals™ Supplier No. NBP210960
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
The NKX3-2 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Cell Line The lysate was created in HEK293T cells, using plasmid ID RC215316 and based on accession number NP_118. The protein contains a C-terminal DDK tag.
For Use With (Application) Western Blot
Quantity 0.1 mg
Storage Buffer RIPA buffer
Purity Protein
Test Specificity Homo sapiens NKX3-2, mRNA.
Content And Storage -80°C. Avoid freeze-thaw cycles.
Protein State Native
For Research Use Only