missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Includes
5mL NHS-Activated Sepharose™ High Performance Medium plus 24 SpinTrap columns
- Antibody or capturing molecule covalently immobilized via free NH2 groups
- Simple coupling, carried out near neutral pH
- Fast and flexible protocol for the capture step, with elution conditions formatted for both electrophoresis and LC-MS analysis workflows
- Bulk media and empty columns allow for batch coupling; use with a standard microcentrifuge
- Easy scale-up to HiTrap™ NHS-activated HP Columns
Specifications
Specifications
| Product Type | NHS HP SpinTrap kit |
| Quantity | Each |
| Includes | Instructions, 5mL NHS activated Sepharose high performance medium in bulk, 24 x empty SpinTrap column |
| Ligand Type | NHS Sepharose HP |
| pH Range | 3 to 12 |
| Particle Size | 34μm |
| Support Type | 6% Highly Cross-linked Spherical Agarose |
| Content And Storage | 4° to 30°C |
| Column Type | Spin Column |
| For Use With (Application) | Coupling of an affinity molecule with a primary Amine |
Safety and Handling
Signal Word
- Danger
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?