missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NEUROG1 Mouse anti-Human, Clone: 1E1H6, Proteintech
Mouse Monoclonal Antibody
1710.33 NOK - 4836.10 NOK
Specifications
| Antigen | NEUROG1 |
|---|---|
| Clone | 1E1H6 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16808715
|
Proteintech
66642-1-IG-20UL |
20 μL |
1710.33 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16898705
|
Proteintech
66642-1-IG-150UL |
150 μL |
4836.10 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Neurogenin 1 is a basic helix-loop-helix (bHLH) proteins are transcription factors involved in determining cell type during development. NeuroG1 is a bHLH protein with dual cell-fate specification roles. It functions during neurogenesis, and it has also been shown to inhibit the differentiation of neural stem cells into astrocytes. NeuroG1 promotes neurogenesis by functioning as a transcriptional activator, yet it inhibits astrocyte differentiation by compartmentalizing the CREB-binding protein transcription complex away from astrocyte differentiation genes and by inhibiting STAT transcription factors essential for gliogenesis.Specifications
| NEUROG1 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| AKA, bHLHa6, Math4C, NEUROD3, NEUROG1, neurogenin 1, NGN, NGN 1, ngn1 | |
| NEUROG1 | |
| IgG1 | |
| Protein G | |
| Antibody |
| 1E1H6 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q92886 | |
| 4762 | |
| NEUROG1 Fusion Protein Ag26438 | |
| Primary | |
| -20°C | |
| NEUROG1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title