missing translation for 'onlineSavingsMsg'
Learn More

NEUROG1 Mouse anti-Human, Clone: 1E1H6, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66642-1-IG-150UL

 View more versions of this product

Product Code. 16898705

  • 4836.10 NOK / 150µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Neurogenin 1 is a basic helix-loop-helix (bHLH) proteins are transcription factors involved in determining cell type during development. NeuroG1 is a bHLH protein with dual cell-fate specification roles. It functions during neurogenesis, and it has also been shown to inhibit the differentiation of neural stem cells into astrocytes. NeuroG1 promotes neurogenesis by functioning as a transcriptional activator, yet it inhibits astrocyte differentiation by compartmentalizing the CREB-binding protein transcription complex away from astrocyte differentiation genes and by inhibiting STAT transcription factors essential for gliogenesis.
Specifications

Specifications

NEUROG1
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q92886
NEUROG1
NEUROG1 Fusion Protein Ag26438
150 μL
Primary
Human
Antibody
IgG1
Western Blot
1E1H6
Unconjugated
NEUROG1
AKA, bHLHa6, Math4C, NEUROD3, NEUROG1, neurogenin 1, NGN, NGN 1, ngn1
Mouse
Protein G
RUO
4762
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado