Learn More
Nav1.6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP3-21269-100ul
 View more versions of this product
Product Code. 18643714
  • ONLINE EXCLUSIVE PRICE
    6370.00 NOK
    6021.75 NOK / 100µL
    Save 348.25 NOK
    5% Off
Description
Nav1.6 Polyclonal antibody specifically detects Nav1.6 in Human samples. It is validated for Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Nav1.6
Polyclonal
Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml
CerIII, hNa6/Scn8a voltage-gated sodium channel, MED, NaCh6, Nav1.6, PN4, sodium channel protein type 8 subunit alpha, Sodium channel protein type VIII subunit alpha, sodium channel, voltage gated, type VIII, alpha polypeptide, sodium channel, voltage gated, type VIII, alpha subunit, Voltage-gated sodium channel subunit alpha Nav1.6
This antibody was developed against Recombinant Protein corresponding to amino acids: EMNNLQISVIRIKKGVAWTKLKVHAFMQAHFKQREADEVKPLDELYEKKANCIANHTGADIHRNGDFQKNGNGTT
100 μg
Neurodegeneration, Neuroscience
6334
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG
Immunofluorescence
Unconjugated
PBS, pH 7.2, 40% glycerol
Rabbit
Affinity purified