Learn More
Fisherbrand™ Natural Rubber Two Hole Stoppers
Product Code. 11547632 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Diameter (Metric) Bottom:
15 mm
17 mm
19 mm
21 mm
23 mm
29 mm
31 mm
33 mm
35 mm
37 mm
39 mm
41 mm
43 mm
45 mm
53 mm
57 mm
62 mm
67 mm
71 mm
75 mm
99 mm
Diameter (Metric) Top:
18 mm
20.3 mm
22.5 mm
24.5 mm
26.5 mm
33 mm
36 mm
38.5 mm
40.5 mm
42.5 mm
44.5 mm
47 mm
49 mm
51 mm
59.5 mm
64 mm
69 mm
74 mm
78 mm
85 mm
109 mm
Unit Size:
Each
Pack of 10
Pack of 2
Pack of 20
Pack of 5
21 product options available for selection
Product selection table with 21 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Bottom Diameter (Metric) Top Unit Size
11547632 29 mm 33 mm Pack of 10
11577622 15 mm 18 mm Pack of 20
11597622 19 mm 22.5 mm Pack of 20
11587622 17 mm 20.3 mm Pack of 20
11507632 21 mm 24.5 mm Pack of 20
11517632 23 mm 26.5 mm Pack of 20
11557632 31 mm 36 mm Pack of 10
11567632 33 mm 38.5 mm Pack of 10
11577632 35 mm 40.5 mm Pack of 10
11587632 37 mm 42.5 mm Pack of 10
11597632 39 mm 44.5 mm Pack of 10
11507642 41 mm 47 mm Pack of 5
11517642 43 mm 49 mm Pack of 5
10549751 45 mm 51 mm Pack of 5
10428972 53 mm 59.5 mm Pack of 2
10559751 57 mm 64 mm Pack of 2
11527642 62 mm 69 mm Pack of 2
11537642 67 mm 74 mm Pack of 2
11547642 71 mm 78 mm Each
11587642 99 mm 109 mm Each
11557642 75 mm 85 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 21 options available.
21 options
Product Code. 11547632
  • ONLINE EXCLUSIVE PRICE
    162.75 NOK
    128.58 NOK / Pack of 10
    Save 34.17 NOK
    20% Off
Estimated Shipment: 23-11-2026
Add to basket
Add to basket
Used for laboratory purposes.
  • Made up of natural rubber
  • Red in color
  • Stoppers with two holes
Specifications
Diameter (Metric) Hole 5 mm
For Use With (Application) For laboratory use
Hardness 46/55 IRHD
Length (Metric) 32 mm
Quantity 10/Pk.
Color Red
Diameter (Metric) Bottom 29 mm
Diameter (Metric) Top 33 mm
Material Natural Rubber
Product Type Tapered Stopper (2 holes)