Learn More
Fisherbrand™ Solid Natural Rubber Stoppers
Product Code. 10192031 Shop All Fisherbrand Products
Click to view available options
Diameter (Metric) Bottom:
6 mm
11 mm
17 mm
23 mm
25 mm
35 mm
57 mm
62 mm
67 mm
105 mm
Diameter (Metric) Top:
8 mm
14 mm
20.3 mm
26.5 mm
28.5 mm
40.5 mm
64 mm
69 mm
74 mm
116.5 mm
Unit Size:
Each
Pack of 10
Pack of 2
Pack of 20
Pack of 50
Product Code. 10192031
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Used for laboratory purposes.
  • Made up of natural rubber
  • Red in color
  • Solid stoppers with no holes
Specifications
For Use With (Application) For laboratory use
Hardness 46/55 IRHD
Length (Metric) 28 mm
Quantity 20/Pk.
Color Red
Diameter (Metric) Bottom 23 mm
Diameter (Metric) Top 26.5 mm
Material Natural Rubber
Product Type Stopper